ResinTech HYDRA-DI Deionizd Water System Cartridge Replacement Kits
Designed to keep your deionized water system running smoothly
  • Easily install and remove cartridges with quarter twist release canister
  • Made in the USA!
Replace your cartridges regularly for best quality and performance. Water purification systems sold separately.
54,800.00 - ₹110,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Capacity (grains) Max Flow Rate (LPM)
Availability Pricing
ResinTech   Replacement Cartridge Kit for Hydra "Slimline" High Purity DI System WW-99285-15 Mfr # HYK-122-S-HP
COMPARE
High Purity Cartridge Replacement Kit for model 99285-11 1800 as CaCO3 1.9
₹54,800.00INR / Each
ResinTech   Replacement Cartridge Kit for Hydra "Big Blue" High Purity DI System WW-99285-16 Mfr # HYK-122-B-HP
COMPARE
High Purity Cartridge Replacement Kit for model 99285-12 4760 as CaCO3 4.7
₹107,000.00INR / Each
ResinTech   Replacement Cartridge Kit for Hydra "Slimline" High Capacity DI System WW-99285-17 Mfr # HYK-122-S-HC
COMPARE
High Capacity Cartridge Replacement Kit for model 99285-13 2160 as CaCO3 1.9
₹55,900.00INR / Each
ResinTech   Replacement Cartridge Kit for Hydra "Big Blue" High Capacity DI System WW-99285-18 Mfr # HYK-122-B-HC
COMPARE
High Capacity Cartridge Replacement Kit for model 99285-14 5710 as CaCO3 4.7
₹110,000.00INR / Each