This product is now discontinued, View Related Products
Red Lion Operator Interface with TFT Color Display
Human machine interface that strikes the perfect balance of advanced features and value!
  • Unit's configuration is stored in nonvolatile Flash memory
  • NEMA 4/IP65 front panel
  • Resistive analog touch-screen
  • Bright TFT display with full color support
  • Both Kadet models are programmed with Red Lion's free Crimson® software
G3 Kadet with TFT Color Display
The Kadet is the perfect solution for applications that require the operator to monitor and control more than just a single device. With multiple serial ports and an Ethernet port, these Kadets can connect to multiple serial and Ethernet devices simultaneously, including PLCs, motor drives, bar code scanners, etc.
Programmed with Red Lion’s free Crimson 2.0 software.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DIN Size On / Off Function PID Function
Availability Pricing
Red Lion G304K200 4.3'' TFT Operator Interface with color display (2 serial ports) WW-68515-96 Mfr # G304K200
COMPARE
Call For Price
Red Lion G307K200 Operator Interface with Color Display; 7" WW-68515-97 Mfr # G307K200