This product is now discontinued, View Related Products
Red Lion N-TRON 100 Series Unmanaged Industrial Ethernet Switches
Increase bandwidth and avoid network collisions
  • Unmanaged internet hub with full IEEE 802.3 compliance
  • High MTBF — greater than 2M hours — ideal for harsh conditions
  • RJ-45 ports support full/half duplex operation and auto-sense duplex, speed, and MDIX
  • Compact footprint conserves space and is DIN-rail mountable
  • ESD and surge protection diodes on all ports
  • Redundant power inputs (10 to 30 VDC) for enhanced reliability
  • Store-and-forward and plug-and-play technology and for ease of use
9 PRODUCT OPTIONS
Compareitem specifications
Max Input Voltage (VDC) Number of Ports Description
Availability Pricing
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 5 Port; SC WW-17702-02 Mfr # 105FX-SC
COMPARE
30 5 N-Tron Unmanaged Industrial Ethernet Switch, 5 Port; SC
Call For Price
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 5 Port; ST WW-17702-04 Mfr # 105FX-ST
COMPARE
30 5 N-Tron Unmanaged Industrial Ethernet Switch, 5 Port; ST
Call For Price
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 6 Port; SC WW-17702-07 Mfr # 106FX2-SC
COMPARE
30 6 N-Tron Unmanaged Industrial Ethernet Switch, 6 Port; SC
Call For Price
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 6 Port; ST WW-17702-09 Mfr # 106FX2-ST
COMPARE
30 6 N-Tron Unmanaged Industrial Ethernet Switch, 6 Port; ST
Call For Price
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 8 Port; TX WW-17702-12 Mfr # 108TX
COMPARE
30 8 N-Tron Unmanaged Industrial Ethernet Switch, 8 Port; TX
Call For Price
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 10 Port; SC WW-17702-14 Mfr # 110FX2-SC
COMPARE
49 10 N-Tron Unmanaged Industrial Ethernet Switch, 10 Port; SC
Call For Price
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 10 Port; ST WW-17702-17 Mfr # 110FX2-ST
COMPARE
49 10 N-Tron Unmanaged Industrial Ethernet Switch, 10 Port; ST
Call For Price
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 12 Port; SC WW-17702-19 Mfr # 112FX4-SC
COMPARE
49 12 N-Tron Unmanaged Industrial Ethernet Switch, 12 Port; SC
Call For Price
Red Lion N-TRON N-Tron Unmanaged Industrial Ethernet Switch, 12 Port; ST WW-17702-21 Mfr # 112FX4-ST
COMPARE
49 12 N-Tron Unmanaged Industrial Ethernet Switch, 12 Port; ST
Call For Price
Showing 1- 9 of 9