Red Lion Data Station Plus Operator Interface with Protocol Translator
A nexus for industrial data collection and management
  • Converts numerous protocols simultaneously
  • Over 200 built-in drivers allow easy data mapping to PLCs, PCs, drives and SCADA systems
  • Supports up to nine protocols simultaneously (with expansion card)
  • DIN rail mounting
  • Operating termperature 32 to 122°F (0 to 50°C)
The Data Station Plus is designed to act as a nexus for industrial data collection and management. With three built-in serial ports and a 10 Base-T/100 Base-TX Ethernet port, the unit performs protocol conversion, allowing disparate devices to communicate seamlessly with one another. The Data Station Plus functions both as a data management platform and as a PFM host.
The Ethernet port supports up to four protocols simultaneously so even Ethernet-to-Ethernet protocol conversion may be performed. The CompactFlash card allows new configuration files to be loaded into the Data Station Plus or use to data log critical information. The USB port may be used for fast file downloads, or to mount the Data Station’s CompactFlash card as an external drive to your PC.
202,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Red Lion DSPLE000 Operator Interface and Protocol Converter WW-68515-98 Mfr # DSPLE000
COMPARE
Operator Interface and Protocol Converter
₹202,000.00INR / Each
Red Lion DSPSX000 Operator Interface and Protocol Converter WW-68515-99 Mfr # DSPSX000
COMPARE
Operator Interface and Protocol Converter
Call For Price
Red Lion PFM1608 Plant Floor Marquee, 26" x 6.6" dimension WW-68516-11 Mfr # PFM1608A
COMPARE
Plant Floor Marquee, 26\" x 6.6\" dimension
Call For Price
Red Lion PFM2412 Plant Floor Marquee, 38.5" x 8.8" dimension WW-68516-12 Mfr # PFM2412A
COMPARE
Plant Floor Marquee, 38.5\" x 8.8\" dimension
Call For Price
Red Lion PFM3212A Plant Floor Marquee, 38.8" x 10.9" dimension WW-68516-13 Mfr # PFM3212A
COMPARE
Plant Floor Marquee, 38.8\" x 10.9\" dimension
Call For Price
Red Lion PFM6412A Plant Floor Marquee, 39.3" x 22.5" dimension WW-68516-14 Mfr # PFM6412A
COMPARE
Plant Floor Marquee, 39.3\" x 22.5\" dimension
Call For Price
Red Lion XCCN0000 Optional CANopen Option Card for Data Station WW-68516-15 Mfr # XCCN0000
COMPARE
Optional CANopen Option Card for Data Station
Call For Price
Red Lion XCDN0000 DeviceNet Option Card WW-68516-16 Mfr # XCDN0000
COMPARE
DeviceNet Option Card
Call For Price
Red Lion XCPBDP00 PROFIBUS Option Card for Data Station WW-68516-17 Mfr # XCPBDP00
COMPARE
PROFIBUS Option Card for Data Station
Call For Price
Red Lion XCRS0000 RS232/485 Option Card WW-68516-18 Mfr # XCRS0000
COMPARE
RS232/485 Option Card
Call For Price
Showing 1- 10 of 10