This product is now discontinued, View Related Products
Cole-Parmer® Essentials Rectangular Tanks with Cover, HDPE and PP
Easy to handle—outside rim flange for easy gripping
  • Rotation-molded in one seamless piece for easy cleaning
  • Chemical resistant
These rectangular tanks are rotation-molded in one seamless piece and have an outside rim flange for strength and ease of handling. Tanks are available in larger capacities and other dimensions; call our Application Specialists for details.
High-Density Polyethylene (HDPE) Tanks have very good chemical resistance and a maximum service temperature of 65°C (150°F). HDPE tanks have a hard, smooth surface that is easy to clean. Tanks are FDA compliant.
Polypropylene (PP) Tanks have very good chemical resistance, excellent stress crack resistance, and a maximum service temperature of 104°C (220°F). PP tanks feature a hard, smooth surface that is easy to clean and is highly resistant to pickling and plating solutions.
Note: Cole-Parmer recommends all rectangular tanks having a dimension of 18" or more, and those used at elevated temperatures or with high specific gravity products, be externally supported. Call our Application Specialists for more information.
31,200.00 - ₹59,900.00INR / Each
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
Compareitem specifications
Material Max Temperature (° F) Volume (Gal)
Availability Pricing
COMPARE
Polypropylene 220 2
Call For Price
COMPARE
Polypropylene 220 6
₹31,200.00INR / Each
COMPARE
Polypropylene 220 6
₹59,200.00INR / Each
COMPARE
Polypropylene 220 7
Call For Price
COMPARE
Polypropylene 220 11
Call For Price
COMPARE
Polypropylene 220 11
₹51,700.00INR / Each
COMPARE
Polypropylene 220 12
Call For Price
COMPARE
Polypropylene 220 15
Call For Price
COMPARE
Polypropylene 220 15
₹59,900.00INR / Each
COMPARE
Polypropylene 220 22
Call For Price
COMPARE
Polypropylene 220 25
Call For Price
COMPARE
Polypropylene 220 30
Call For Price
COMPARE
Polypropylene 220 45
Call For Price
COMPARE
Polypropylene 220 60
Call For Price
COMPARE
Polypropylene 220 90
Call For Price
COMPARE
Polypropylene 220 140
Call For Price
COMPARE
LLDPE 150 2
Call For Price
COMPARE
HDPE 150 6
Call For Price
COMPARE
HDPE 150 6
₹39,200.00INR / Each
COMPARE
HDPE 150 7
Call For Price
Showing 1 - 20 of 32