Rechargeable Ultraviolet Lamps
Portable design is great for use in the field
Choose from single- or dual-wavelength lamps; all include 6-watt tube(s). Single wavelength lamps let you use both tubes simultaneously to achieve intensities twice as high as similar units. Dual-wavelength lamps let you choose from either 254 or 365 nm light.
Rechargeable ultraviolet lamps are powered by the built-in rechargeable battery or included AC/recharger adapter (60 Hz for 115 VAC; 50 Hz for 230 VAC). The optional cigarette lighter adapter (sold separately) lets you operate UV lamps on a 12 VDC power source.The dual-wavelength models require one replacement tube for each wavelength. Lamps measure 3"W x 9-3/4"H x 5-1/2"D. Shpg wt 4.0 lb (1.9 kg).
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC)
Availability Pricing
Analytik Jena Rechargeable ultraviolet lamp; 365 nm wavelength, 220 VAC/50 Hz WW-97603-05 Mfr # 95-0186-02
COMPARE
50 230