VistaLab™ Reagent Reservoirs, Polystyrene
Deep-well design allows you to pipette even small amounts of reagent
  • Trough-within-a-trough design to recover the “last few drops”
  • Pour-off spouts at every corner
  • Constructed of pyrogen-free polystyrene
These recyclable reagent reservoirs are available in a variety of volumes and packaging configurations. Constructed of pyrogen-free polystyrene, they will preserve the integrity of your sample. Well-marked graduated increments on the inside wall (all sizes except 100 mL) help you visualize reagent levels and minimize waste.
Wide base and sturdy construction provide stability to avoid spills. Unique trough design allows even small amounts of liquid to be drawn through all pipette tips. Trimmed base corners and distinct pour-off spouts make it easy to empty the reservoir without spills.
Order the reusable, nontoxic gel cooler to preserve the integrity of reagents and extend working time with full reservoir cooling. Custom fit for the 100-mL reagent reservoirs, the cooler will maintain your reagent at <41°F (5°C) for up to two hours.
16,800.00 - ₹21,900.00INR / Case of 100
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Number Of Channels Sterile
Availability Pricing
VistaLab Reagent Reservoir, Polystyrene, Sterile, 25 mL; 100/Cs WW-06198-70 Mfr # 3054-1002
COMPARE
25 1 Yes
₹19,500.00INR / Case of 100
VistaLab Reagent Reservoir, Polystyrene, Sterile, 25 mL, Divided; 100/Cs WW-06198-71 Mfr # 3054-1004
COMPARE
25 2 Yes
₹20,200.00INR / Case of 100
VistaLab Reagent Reservoir, Polystyrene, Sterile, 50 mL; 100/Cs WW-06198-72 Mfr # 3054-1006
COMPARE
50 1 Yes
₹19,500.00INR / Case of 100
VistaLab Reagent Reservoir, Polystyrene, Sterile, 100 mL; 100/Cs WW-06198-73 Mfr # 3054-1000
COMPARE
100 1 Yes
₹19,500.00INR / Case of 100
VistaLab Reagent Reservoir, Polystyrene, Non-Sterile, 25 mL; 100/Cs WW-06198-74 Mfr # 4054-1002
COMPARE
25 1 No
₹17,500.00INR / Case of 100
VistaLab Reagent Reservoir, Polystyrene, Non-Sterile, 25 mL, Divided; 100/Cs WW-06198-75 Mfr # 4054-1004
COMPARE
25 2 No
₹17,500.00INR / Case of 100
VistaLab Reagent Reservoir, Polystyrene, Non-Sterile, 50 mL; 100/Cs WW-06198-76 Mfr # 4054-1006
COMPARE
50 1 No
₹16,800.00INR / Case of 100
VistaLab Reagent Reservoir, Polystyrene, Non-Sterile, 100 mL; 100/Cs WW-06198-77 Mfr # 4054-1000
COMPARE
100 1 No
₹16,800.00INR / Case of 100
VistaLab Reagent Reservoir, Polystyrene, Sterile and Individually Wrapped, 25 mL; 50/Cs WW-06198-78 Mfr # 3054-1003
COMPARE
25 1 Yes
₹21,900.00INR / Case of 50
VistaLab Reagent Reservoir, Polystyrene, Sterile and Individually Wrapped, 25 mL, Divided; 50/Cs WW-06198-79 Mfr # 3054-1005
COMPARE
25 2 Yes
₹21,000.00INR / Case of 50
VistaLab Reagent Reservoir, Polystyrene, Sterile and Individually Wrapped, 50 mL; 50/Cs WW-06198-80 Mfr # 3054-1007
COMPARE
50 1 Yes
₹21,900.00INR / Case of 50
VistaLab Reagent Reservoir, Polystyrene, Sterile and Individually Wrapped, 100 mL; 50/Cs WW-06198-81 Mfr # 3054-1001
COMPARE
100 1 Yes
₹21,900.00INR / Case of 50
Showing 1- 12 of 12