Raytek CM-Series Compact Infrared Sensors
The Raytek CM Sensor is the ideal solution for both OEM and end-user applications – compact, powerful, affordable and easy to install.
  • Replaces traditional contact probes with a scaleable 0-5 V, type J or K output
  • Wide temperature range: -4ºF to 932ºF (-20 to 500ºC) °
  • Rugged 13:1optics at 90% energy
  • Rugged IP 65 (NEMA-4) stainless steel housing
  • Sensor status/self-diagnostic indicator LED
The Raytek CM Sensor is designed for continuous temperature monitoring in a broad range of manufacturing processes and OEM applications. The sensor provides the advantages of infrared temperature measurement in a powerful, compact, integrated sensor. The CM sensor can easily replace traditional contact probes with a J-type or K-type thermocouple output, or with a user scaleable 0-5 volt output. The sensors onboard electronics are protected by a rugged IP 65 (NEMA-4) stainless steel housing, allowing the sensor to function in ambient temperatures up to 160°F (70°C) without cooling. The built-in sensor health LED provides a convenient on-line indicator of the sensor's operating status and aids in troubleshooting initial sensor set up. Although small in size, the CM is loaded with advanced features including; RS232 digital interface, 24 volt DC alarm, peak hold, valley hold, variable averaging signal processing, adjustable target emissivity settings and adjustable window transmissivity. DataTemp Multidrop software is includes with each sensor allowing the user to configure all programmable sensor variables.
83,400.00 - ₹93,600.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° C) Description
Availability Pricing
Raytek Compact CM-Series Infrared Sensor Type-J Thermocouple Probe Output with 3' Cable WW-39660-00 Mfr # RAYCMLTJ
COMPARE
932 -20 Compact CM-Series Infrared Sensor Type-J Thermocouple Probe Output with 3' Cable
₹83,400.00INR / Each
Raytek Compact CM-Series Infrared Sensor Type-J Thermocouple Probe Output with 10' Cable WW-39660-01 Mfr # RAYCMLTJ3
COMPARE
932 -20 Compact CM-Series Infrared Sensor Type-J Thermocouple Probe Output with 10' Cable
₹93,600.00INR / Each
Raytek Compact CM-Series Infrared Sensor Type-K Thermocouple Probe Output with 3' Cable WW-39660-02 Mfr # RAYCMLTK
COMPARE
932 -20 Compact CM-Series Infrared Sensor Type-K Thermocouple Probe Output with 3' Cable
₹83,400.00INR / Each
Raytek Compact CM-Series Infrared Sensor Type-K Thermocouple Probe Output with 10' Cable WW-39660-03 Mfr # RAYCMLTK3
COMPARE
932 -20 Compact CM-Series Infrared Sensor Type-K Thermocouple Probe Output with 10' Cable
₹93,600.00INR / Each
Raytek Compact CM-Series Infrared Sensor Voltage Output with 3' Cable WW-39660-04 Mfr # RAYCMLTV
COMPARE
932 -20 Compact CM-Series Infrared Sensor Voltage Output with 3' Cable
₹83,400.00INR / Each
Raytek Compact CM-Series Infrared Sensor Voltage Output with 10' Cable WW-39660-05 Mfr # RAYCMLTV3
COMPARE
932 -20 Compact CM-Series Infrared Sensor Voltage Output with 10' Cable
₹93,600.00INR / Each