QuadMag Genie Multiposition Stirrer 62313
Durable, reliable multiposition mixing
  • Ideal for use in cold rooms and incubators
  • Stir up to 2 L per position
This four-position stirrer features powerful magnets that will pull stir bars into position
and remain coupled even under the most demanding stirring conditions. The durable,
spill-resistant ABS/polycarbonate housing is designed to direct spills away from the internal
components. Internal ramping feature will gradually increase speed to the set point to
minimize splashing and improve control of the stir bar. Control speed with the graduated
analog dial
254,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-536 Digital Quadmag Multiposition Stirrer, 120V WW-86575-23 Mfr # SI-5236
COMPARE
120
₹254,000.00INR / Each
Scientific Industries Genie SI-5256 Digital Quadmag Mulriposition Stirrer, 230V WW-86575-24 Mfr # SI-5256
COMPARE
230
₹254,000.00INR / Each