Cole-Parmer® Essentials Safety Coated Sample Jars, Clear Glass
Will not shatter
  • Reduce breakage with safety-coated design
  • Chemical-resistant
Impact-resistant, clear plastisol coating makes these jars virtually shatterproof—reduces breakage and prevents accidents in the lab. Chemical-resistant, PTFE-lined screw caps provide a positive, liquid-tight seal even after repeated openings.
21,900.00 - ₹46,600.00INR / Case of 24
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Cole-Parmer Essentials 7533TW Safety Coated Sample Jar, Clear Glass, 250mL; 24/CS WW-34604-10 Mfr # GLC-02240
COMPARE
250 70 mm Glass
₹40,500.00INR / Case of 24
Cole-Parmer Essentials 7785TW Safety Coated Sample Jar, Clear Glass, 500mL; 24/CS WW-34604-20 Mfr # GLC-02236
COMPARE
500 70 mm Glass
₹46,600.00INR / Case of 24
Cole-Parmer Essentials 7772TW Safety Coated Sample Jar, Clear Glass, 1000mL; 12/CS WW-34604-30 Mfr # GLC-02245
COMPARE
1000 70 mm Glass
₹29,700.00INR / Case of 12
Cole-Parmer Essentials GLC05729 Safety-coated Sample Jars, Standard Rounded, 2000mL; 6/CS WW-34604-40 Mfr # GLC-05729
COMPARE
2000 83 mm Glass
₹21,900.00INR / Case of 6