Cole-Parmer® Essentials Pre-Cleaned Wide-Mouth Jars, Clear Glass
Save time—no need to prewash bottles
  • Pre-cleaned by vacuum and ionization
  • Individually sealed for long term storage with no contamination
The jars are individually sealed for long-term storage with no contamination. Available with PTFE-lined caps.
16,600.00 - ₹24,400.00INR / Case of 12
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottle Color Capacity (mL) Material
Availability Pricing
Cole-Parmer Essentials 2S01 QGTV Pre-Cleaned Wide-Mouth Jar, Clear Glass, 45mL; 48/CS WW-34608-00 Mfr # GLC-01588
COMPARE
Clear 45 Glass
₹24,400.00INR / Case of 48
Cole-Parmer Essentials 2S02 QGTV Pre-Cleaned Wide-Mouth Jar, Clear Glass, 60mL; 24/CS WW-34608-10 Mfr # GLC-01613
COMPARE
Clear 60 Glass
₹16,600.00INR / Case of 24
Cole-Parmer Essentials 2S04 QGTV Pre-Cleaned Wide-Mouth Jar, Clear Glass, 125mL; 24/CS WW-34608-20 Mfr # GLC-01636
COMPARE
Clear 125 Glass
₹18,100.00INR / Case of 24
Cole-Parmer Essentials 2S08 QGTV Pre-Cleaned Wide-Mouth Jar, Clear Glass, 250mL; 24/CS WW-34608-30 Mfr # GLC-01674
COMPARE
Clear 250 Glass
₹23,300.00INR / Case of 24
Cole-Parmer Essentials 2S16 QGTV Pre-Cleaned Wide-Mouth Jar, Clear Glass, 500mL; 12/CS WW-34608-40 Mfr # GLC-01694
COMPARE
Clear 500 Glass
₹19,300.00INR / Case of 12
Cole-Parmer Essentials 2S32 QGTV Pre-Cleaned Wide-Mouth Jar, Clear Glass, 1000mL; 12/CS WW-34608-50 Mfr # GLC-01723
COMPARE
Clear 1000 Glass
₹21,800.00INR / Case of 12