This product is now discontinued, View Related Products
Qorpak Kaptclean Sample Jars
No precleaning requred
  • Ideal for environmental sampling
  • Chemical-resistant PTFE-lined screw cap
These wide-mouth Kaptclean® glass jars are pre-cleaned for semivolatiles, pesticides, PCBs, and metals—ideal for environmental sampling. Manufactured from USP Type III soda-lime glass. Jars come with chemical-resistant PTFE liner inserted and screw cap attached. Sample jars include a certificate of analysis.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Cap Material Material
Availability Pricing
COMPARE
120 PTFE-lined Glass
Call For Price
COMPARE
250 PTFE-lined Glass
Call For Price
COMPARE
500 PTFE-lined Glass
Call For Price
COMPARE
960 PTFE-lined Glass