This product is now discontinued, View Related Products
QLA Vessels Compatible with Agilent 708/709-DS Dissolution Apparatus
QLA vessels provide more precise and consistent dissolution results
  • Unique serial number is laser marked on plastic rim of each vessel
  • Plastic rims help minimize glass breakage
  • 1000 mL capacity
Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle.
40,900.00 - ₹55,500.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Bottle Color Material Special Features
Availability Pricing
QLA Standard Vessel with Plastic Rim Compatible with Agilent 708/709-DS Baths, 1000 mL, Clear Glass; Serialized WW-41306-20 Mfr # GLA900-70XDS
COMPARE
Clear Glass Standard with plastic rim
Call For Price
QLA Standard Vessel with Plastic Rim Compatible with Agilent 708/709-DS Baths, 1000 mL, Amber Glass; Serialized WW-41306-21 Mfr # GLA900-A70XDS
COMPARE
Amber Glass Standard with plastic rim
₹40,900.00INR / Each
QLA PEAK Vessel with Plastic Rim Compatible with Agilent 708/709-DS Baths, 1000 mL, Clear Glass; Serialized WW-41306-48 Mfr # GLA9PK-70XDS
COMPARE
Clear Glass PEAK style with plastic rim
₹44,100.00INR / Each
QLA PEAK Vessel with Plastic Rim Compatible with Agilent 708/709-DS Baths, 1000 mL, Amber Glass; Serialized WW-41306-49 Mfr # GLA9PK-A70XDS
COMPARE
Amber Glass PEAK style with plastic rim
₹55,500.00INR / Each