QLA Dissolution Filter Tips Compatible with Agilent and Legacy VanKel Baths
Achieve reproducibility and consistency from test to test with QLA dissolution filters
  • Ideal for process involving large diameter sample probes
  • Keep contaminates out of dissolution samples
  • Guaranteed free of chromium and heavy metal
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filter feature Ultra High Molecular Weight (UHMW) polyethylene membranes and polypropylene housing.
54,600.00INR / Pkg of 1000
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (micron)
Availability Pricing
QLA Filter Tips Compatible with Agilent and VanKel Dissolution Apparatus, 2 µm, 1/8" ID; 1000/PK WW-41309-94 Mfr # FILTIP-02
COMPARE
2
₹54,600.00INR / Pkg of 1000
QLA Filter Tips Compatible with Agilent and VanKel Dissolution Apparatus, 5 µm, 1/8" ID; 1000/PK WW-41309-95 Mfr # FILTIP-05
COMPARE
5
₹54,600.00INR / Pkg of 1000
QLA Filter Tips Compatible with Agilent and VanKel Dissolution Apparatus, 10 µm, 1/8" ID; 1000/PK WW-41309-96 Mfr # FILTIP-10
COMPARE
10
₹54,600.00INR / Pkg of 1000
QLA Filter Tips Compatible with Agilent and VanKel Dissolution Apparatus, 20 µm, 1/8" ID; 1000/PK WW-41309-97 Mfr # FILTIP-20
COMPARE
20
₹54,600.00INR / Pkg of 1000
QLA Filter Tips Compatible with Agilent and VanKel Dissolution Apparatus, 45 µm, 1/8" ID; 1000/PK WW-41309-99 Mfr # FILTIP-45
COMPARE
45
₹54,600.00INR / Pkg of 1000