PyrexBrand 6140 Glass Funnels
Permits accurate fitting of the filter paper
  • Beaded edge for added strength
  • 60° plain angle, long stem
These funnels are fabricated from blanks formed to a 60 degree angle, which permits accurate fitting of the filter paper, thus reducing filtering time. Manufactured with a beaded edge for additional strength and stems are constructed with heavy wall tubing.
10,000.00 - ₹29,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Filter Paper Diameter (mm) Material
Availability Pricing
Pyrex 6140-50 Glass Funnel, 50 mm top dia, 28 mL, 6/PK WW-34560-50 Mfr # 6140-50
COMPARE
28 50 Glass
₹20,400.00INR / Each
Pyrex 6140-65 Glass Funnel, 65 mm top dia, 62 mL, 6/PK WW-34560-51 Mfr # 6140-65
COMPARE
62 65 Glass
₹13,900.00INR / Each
Pyrex 6140-75 Glass Funnel, 75 mm top dia, 94 mL, 6/PK WW-34560-52 Mfr # 6140-75
COMPARE
94 75 Glass
₹15,000.00INR / Each
Pyrex 6140-100 Glass Funnel, 100 mm top dia, 225 mL, 3/PK WW-34560-53 Mfr # 6140-100
COMPARE
235 100 Glass
₹10,000.00INR / Each
Pyrex 6140-75 Glass Funnel, 75 mm top dia, 94 mL, 12/CS WW-34560-62 Mfr # 6140-75
COMPARE
94 75 Glass
₹29,900.00INR / Case of 12
Pyrex 6140-100 Glass Funnel, 100 mm top dia, 225 mL, 6/CS WW-34560-63 Mfr # 6140-100
COMPARE
235 100 Glass
₹20,000.00INR / Case of 6