PYREX® Brand 5660 Lifetime Red™ Class A Volumetric Flasks
No more worn off graduations
  • Get permanent legibility and accuracy with etched red graduation
  • Volumetric flasks are constructed of borosilicate glass
These PYREX® Brand 5660 Class A volumetric flasks feature a fine line capacity mark etched through a narrow red band into clear glass, making a filler unnecessary and providing high legibility, accuracy, and permanence.
The strength of these flasks has been increased appreciably through machine-blown bodies that feature sealed heavy-tubing necks.
Flasks are calibrated "To Contain" and meet Class A tolerances as established by ASTM E- 694 for volumetric ware, ASTM E-542 for calibration of volumetric ware, and ASTM E-288 for volumetric flasks.
51,400.00 - ₹128,000.00INR / Pkg of 6
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stopper Number Accepted Tolerance (mL)
Availability Pricing
Pyrex Brand 5660 Lifetime Red™ Class A Volumetric Flask; 100 mL; 6/PK WW-34562-03 Mfr # 5660-100
COMPARE
100 13 ±0.08
₹51,400.00INR / Pkg of 6
Pyrex Brand 5660 Lifetime Red™ Class A Volumetric Flask; 25 mL; 12/CS WW-34562-21 Mfr # 5660-25
COMPARE
25 9 ±0.03
₹90,300.00INR / Case of 12
Pyrex Brand 5660 Lifetime Red™ Class A Volumetric Flask; 50 mL; 12/CS WW-34562-22 Mfr # 5660-50
COMPARE
50 9 ±0.05
₹96,300.00INR / Case of 12
Pyrex Brand 5660 Lifetime Red™ Class A Volumetric Flask; 100 mL; 12/CS WW-34562-23 Mfr # 5660-100
COMPARE
100 13 ±0.08
₹103,000.00INR / Case of 12
Pyrex Brand 5660 Lifetime Red™ Class A Volumetric Flask; 250 mL; 12/CS WW-34562-25 Mfr # 5660-250
COMPARE
250 16 ±0.12
₹128,000.00INR / Case of 12
Pyrex Brand 5660 Lifetime Red™ Class A Volumetric Flask; 1000 mL; 6/CS WW-34562-27 Mfr # 5660-1L
COMPARE
1000 22 ±0.3
₹96,300.00INR / Case of 6
Pyrex Brand 5660 Lifetime Red™ Class A Volumetric Flask; 2000 mL; 4/CS WW-34562-28 Mfr # 5660-2L
COMPARE
2000 27 ±0.05
₹93,900.00INR / Case of 4
Showing 1- 7 of 7