PVC Hose and Cuffs
This PVC hose is ideal for transfer, vacuum, and high flow rate applications. Heavy-duty, flexible construction resists moisture and chemical corrosion. "Spring-back" coil provides optimum crush resistance. maximum operating pressure is 44 psi at 72°>F; temperature range is 30 to 180°>F (-1 to 82°>C). Order PVC cuffs for simple, air tight hose installation.
5,300.00 - ₹49,400.00INR / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
1 1 13/64
₹5,300.00INR / Pkg of 1
COMPARE
1 1 13/64
₹49,400.00INR / Pkg of 1