Chromatography Research Supplies Puresep™ T Septa

Designed for enhanced bond and durability
  • Silicone-based septum is faced on one side with a double layer of polyimide and PTFE
PURESEP-T™ septa are made of PTFE and polyimide films on silicone rubber bases. PTFE gives maximum inertness and low bleed. Maximum temperature: 437°F (225°C).
8,800.00 - ₹10,500.00INR / Pkg of 50

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Chromatography Research Supplies Puresep™ T Septa, 1/4" (6.0 mm); 50/Pk WW-01994-20 Mfr # 230665
COMPARE
Septa, 1/4\" (6.0 mm); 50/Pack
₹8,800.00INR / Pkg of 50
Chromatography Research Supplies Puresep™ T Septa, 3/8" (9.5 mm); 50/P WW-01994-21 Mfr # 230675
COMPARE
Septa, 3/8\" (9.5 mm); 50/Pack
₹8,800.00INR / Pkg of 50
Chromatography Research Supplies Puresep™ T Septa, 7/16" (11.0 mm); 50/Pk WW-01994-22 Mfr # 230685
COMPARE
Septa, 7/16\" (11.0 mm); 50/pack
₹9,900.00INR / Pkg of 50
Chromatography Research Supplies Puresep™ T Septa, 1/2" (12.5 mm); 50/Pk WW-01994-23 Mfr # 230695
COMPARE
Septa, 1/2\" (12.5 mm); 50/pack
₹10,500.00INR / Pkg of 50
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat