This product is now discontinued, View Related Products
PRP-X300 Ion Exclusion Columns
Efficient separations with durable media
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene) stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-X300 Ion Exclusion Columns contain a partially sulfonated poly (styrene-divinylbenzene) media. Exhibit a high degree of both reverse phase and ion exclusion characteristics; separations are based on charge as well as relative hydrophobicity. Ideal for analysis of carboxylic acids and alcohols—no elevated temperature required. Exchange capacity is 0.17 mEq/g.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Column Particle Size (µm) Wetted Materials Tubing Inside Diameter (ID) (mm)
Availability Pricing
Hamilton 79464 PRP-X300 Ion Exclusion Column, Fast Analytical, 150x4.1mm ID; 7um WW-01944-00 Mfr # 79464
COMPARE
7
Call For Price
Hamilton 79465 PRP-X300 Ion Exclusion Column, Analytical, 250x4.1mm ID; 7um WW-01944-02 Mfr # 79465
COMPARE
8