Probes for Q55 Sonicator Ultrasonic Processor

Increase the versatility of your Qsonica Q55 Sonicator with additional probes
  • Choose probe size based on sample size
53,600.00 - ₹109,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Min Sample Size (mL) Tip Diameter (in)
Availability Pricing
Qsonica 4423 Q55 Sonicator Microprobe, 5/64", 0.2 - 5 mL WW-04712-53 Mfr # 4423
COMPARE
5 0.2 0.0781
₹59,500.00INR / Each
Qsonica 4422 Q55 Sonicator Microprobe, 1/8", 0.5 - 15 mL WW-04712-54 Mfr # 4422
COMPARE
15 0.5 0.125
₹53,600.00INR / Each
Qsonica 4435 Q55 Sonicator Microprobe, 1/4", 10 - 50 mL WW-04712-55 Mfr # 4435
COMPARE
50 10 0.25
₹53,600.00INR / Each
Qsonica 4219 1/2" SOLID PROBE* WW-04712-67 Mfr # 4219
COMPARE
₹109,000.00INR / Each
Qsonica 4209 1" SOLID PROBE* WW-04712-71 Mfr # 4209
COMPARE
₹109,000.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat