This product is now discontinued, View Related Products
Presterilized PES Filter Capsules
Improved wetability for critical processes
The capsule contains two layers of polyethersulfone (PES) membrane, which is inherently hydrophilic, low in extractables, and has excellent flow rates. The membrane also exhibits low protein binding characteristics making it ideal for use with biologically sensitive solutions. The device is supplied presterilized and has a vent to evacuate any air to allow fast filtration. Polypropylene housing. Some capsules available with filling bell for cleanliness assurance during filling.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Connections Effective Filtration Area (cm²) Pore Size (micron)
Availability Pricing
ZenPure Filter Capsule PES, 0.2um, 550 cm2, Sterile, 1/4" to 1/2" barb WW-49701-44 Mfr # MKPS020020C2H2H-ETO
COMPARE
1/4\" to 1/2\" hose barb 400 0.2/0.2
Call For Price
ZenPure Filter Capsule PES, 0.65um/0.45um, sterile, 1/4" to 3/8" barb connection WW-49701-46 Mfr # MZPS065045C2H2H-GIO
COMPARE
1/4\" to 1/2\" hose barb 400 0.65/0.45
Call For Price
ZenPure Filter Capsule PES, 0.2um, 550 cm2, sterile, 1/4" to 1/2" barb, filling bell WW-49701-48 Mfr # MZPS020020C2H2H-FC-GIO
COMPARE
1/4\" to 1/2\" hose barb 400 0.2/0.2
Call For Price
ZenPure Filter Capsule PES, 0.8um/0.2um, 380 cm2 filtration area, sterile, 1/4" to 3/8" WW-49701-50 Mfr # MZPS080020B2H2H-FC-GIO
COMPARE
1/4\" to 1/2\" hose barb 300 0.8/0.2
Call For Price
ZenPure Filter Capsule PES, 0.65um/0.45um, 550 cm2 filtration area, sterile, 1/4" to 3/8" WW-49701-52 Mfr # MZPS065045C2H2H-FC-GIO
COMPARE
1/4\" to 1/2\" hose barb 400 0.65/0.45
Call For Price
ZenPure Filter Capsule PES, 0.2um, 1100 cm2, sterile, 1/4" to 3/8" barb WW-49701-54 Mfr # SZLS020020S2H2H-GIO
COMPARE
1/4\" to 1/2\" hose barb 1380 0.2/0.2
Call For Price
ZenPure Filter Capsule PES, 0.2um, 1100 cm2, 1/4" to 3/8" barb WW-49701-58 Mfr # SZLS020020S2H2H-FB-ETO
COMPARE
1/4\" to 1/2\" hose barb 1380 0.2/0.2
Call For Price
ZenPure Filter Capsule PES, 0.8um/0.2um, 760 cm2, sterile, 1/4" to 3/8" barb WW-49701-60 Mfr # SZLS080020H2H2H-FC-GIO
COMPARE
1/4\" to 1/2\" hose barb 720 0.8/0.2
Call For Price
ZenPure Filter Capsule PES, 0.2um, 2200 cm2, sterile, 3/8" to 1/2" barb Sku4970164 WW-49701-64 Mfr # SZLS020020L3H3H-GIO
COMPARE
3/8\" hose barb 1300 0.2/0.2
Call For Price
ZenPure Filter Capsule PES, 0.65um/0.45um, 2200 cm2, sterile, 1/4" to 3/8" barb WW-49701-66 Mfr # SZLS065045L3H3H-GIO
COMPARE
3/8\" hose barb 2600 0.65/0.45
Call For Price
ZenPure Filter Capsule PES, 0.2um, 2200 cm2, sterile, 3/8" to 1/2" barb Sku4970168 WW-49701-68 Mfr # SZLS020020L3H3H-FC-GIO
COMPARE
3/8\" hose barb 2600 0.2/0.2
Call For Price
ZenPure Filter Capsule PES, 0.8um/0.2um, 1140 cm2, sterile, 1/4" to 3/8" barb WW-49701-70 Mfr # SZLS080020S2H2H-FC-GIO
COMPARE
3/8\" hose barb 1380 0.8/0.2
Call For Price
Showing 1- 12 of 12