This product is now discontinued, View Related Products
Pressure Meter with Formatted Data Logging SD Card
Measure a variety of pressure ranges with a single meter
  • Settable parameters of date, time, auto power off, beep sound, sampling time, decimal point or comma decimal division
  • Data logging sampling time settable from 1 second to 9 hours
  • Minimum and maximum recallable readings
This versatile meter, along with the interchangeable transducers, provides a great solution for pressure measurement requirements in HVAC/R, industrial, and process applications. The transducers connect to the meter via a 4-pin M12 electronic connection with a 3-ft (0.9-m) long cable. The 2 GB SD card stores data in Excel® format for logging and downloading onto your computer for record keeping.
Note: Interchangeable pressure tranducers are required for use.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
General Tools & Instruments PS10 Pressure Transmitter for 68602-20, 0 to 145 psi WW-68602-26 Mfr # PS10