Precolumn filters offer secure protection for analytical columns
Economical protection for larger columns and injections
Feature 10-32 coned ports for connection to 1/16" OD tubing
Replaceable stainless steel (SS) and PEEK® frits
Economical precolumn filters are designed to protect columns by filtering out particulate matter originating from the sample or from rotor seal wear. Filters feature 10-32 coned ports compatible with 1/16" outer diameter tubing.