Aven Tools Precision Tweezers, Stainless Steel
Ideal for micro electronics and precision assembly
  • Lifetime warranty
These finely crafted stainless steel tweezers feature hand finished points for accuracy and excellent tip hardness. Anti-glare finish reduces eye strain. The tweezers are also anti-magnetic, anti-acid, and acid resistant.
1,200.00 - ₹3,000.00INR / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in) Style
Availability Pricing
Aven Tools Tweezers, Stainless Steel, Straight, Strong Precision Tips, 5"L WW-07287-01 Mfr # 18013USA
COMPARE
Stainless Steel 5 Straight, strong precision tips
₹1,200.00INR / Pkg of 1
Aven Tools Tweezers, Stainless Steel, Straight, Thick, Flat, Strong Tips, 4-3/4"L WW-07287-02 Mfr # 18032USA
COMPARE
Stainless Steel 4 3/4 Straight, Thick, Flat, Strong Tips
₹1,200.00INR / Pkg of 1
Aven Tools Tweezers, Stainless Steel, Serrated Straight Tips, 4.75" L WW-07287-03 Mfr # 18037USA
COMPARE
Stainless Steel 4 3/4 Straight, Thick, Flat, Strong Tips
₹1,400.00INR / Pkg of 1
Aven Tools Tweezers, Stainless Steel, Straight, Narrow, Long Fine Tips, 5-1/2"L WW-07287-04 Mfr # 18040USA
COMPARE
Stainless Steel 5 1/2 Straight, Narrow, Long Fine Tips
₹3,000.00INR / Pkg of 1
Aven Tools Tweezers, Stainless Steel, Tapered, Flat, Tips, 4-3/4"L WW-07287-05 Mfr # 18049USA
COMPARE
Stainless Steel 4 3/4 Tapered, Flat, Tips
₹3,000.00INR / Pkg of 1
Aven Tools Tweezers, Stainless Steel, Straight, Ultra Fine Sharp Tips, 4-1/4"L WW-07287-06 Mfr # 18056USA
COMPARE
Stainless Steel 4 1/4 Straight, Ultra Fine Sharp Tips
₹1,200.00INR / Pkg of 1
Aven Tools Tweezers, Stainless Steel, Tapered, Ultra Fine Tips, 4-1/4"L WW-07287-07 Mfr # 18062USA
COMPARE
Stainless Steel 4 1/4 Curved, Fine Tips
₹3,000.00INR / Pkg of 1
Aven Tools Tweezers, Stainless Steel, Angled, Ultra Fine Tips, 4-1/4"L WW-07287-08 Mfr # 18066USA
COMPARE
Stainless Steel 4 1/4 Angled, Ultra Fine Tips
₹1,300.00INR / Pkg of 1
Showing 1- 8 of 8