Precision Tweezers, Plastic
Ideal for handling semiconductors, silicone chips, and delicate wafers. Static dissipative for use with sensitive components where ESD damage is a concern. Temperature resistant up to 220°C; tips do not spread under pressure.
5,800.00 - ₹7,000.00INR / Pkg of 12
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in) Sterile
Availability Pricing
Aven Tools Tweezers, curved, Smooth Rounded Tips 2AB WW-07290-06 Mfr # 18.514
COMPARE
Plastic 4 1/2 No
₹7,000.00INR / Pkg of 12
Aven Tools Tweezers, curved, Very Fine and Smooth Tips 7-STD WW-07290-08 Mfr # 18.523
COMPARE
Plastic 4 1/2 No
₹5,800.00INR / Pkg of 12