Advantech Precision Sieves, Nickel Mesh, 3" dia
Precision sieves feature electroformed nickel mesh. Sieves meet ASTM E161 and ISO specifications. Sieves measure 3" ID x 1-1/4" H.
193,000.00 - ₹332,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (InchShort) Material Sieve Opening (mm)
Availability Pricing
Advantech L3-M3 Precision Sieves, Nickel Mesh, 0.003 mm Sieve Opening, 3" dia WW-59948-83 Mfr # L3-M3
COMPARE
1.25 Acrylic frame, electroformed nickel wire 0.003
₹332,000.00INR / Each
Advantech L3-M5 Precision Sieves, Nickel Mesh, 0.005 mm Sieve Opening, 3" dia WW-59948-84 Mfr # L3-M5
COMPARE
1.25 Acrylic frame, electroformed nickel wire 0.005
₹332,000.00INR / Each
Advantech L3-M10 Precision Sieves, Nickel Mesh, 0.01 mm Sieve Opening, 3" dia WW-59948-85 Mfr # L3-M10
COMPARE
1.25 Acrylic frame, electroformed nickel wire 0.01
₹241,000.00INR / Each
Advantech L3-M15 Precision Sieves, Nickel Mesh, 0.015 mm Sieve Opening, 3" dia WW-59948-86 Mfr # L3-M15
COMPARE
1.25 Acrylic frame, electroformed nickel wire 0.015
₹241,000.00INR / Each
Advantech L3-M20 Precision Sieves, Nickel Mesh, 0.02 mm Sieve Opening, 3" dia WW-59948-87 Mfr # L3-M20
COMPARE
1.25 Acrylic frame, electroformed nickel wire 0.02
₹241,000.00INR / Each
Advantech L3-M100 Precision Sieves, Nickel Mesh, 0.1 mm Sieve Opening, 3" dia WW-59948-91 Mfr # L3-M100
COMPARE
1.25 Acrylic frame, electroformed nickel wire 0.1
₹193,000.00INR / Each