DWK Wheaton® Potter-Elvehjem Safe-Grind Tissue Grinders
Plastic-coated glass mortar increases safety in the lab
  • PTFE-tipped stainless steel rod fits overhead stirrer collets or chucks designed for 0.25" shafts
The heavy plastisol coating protects the mortar from being scratched or shattered, and provides the user with a sure grip. Stainless steel rod is 0.25" (6.3 mm) in diameter. Grinding chamber clearance is 0.004" to 0.006". Unit can be autoclaved.
54,800.00 - ₹74,300.00INR / Case of 2
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Overall Length (mm)
Availability Pricing
DWK Life Sciences (Wheaton) 358003 Potter-Elvehjem Safe-Grind Tissue Grinders, 2 mL WW-04468-26 Mfr # 358003
COMPARE
2 Glass 203
₹54,800.00INR / Case of 2
DWK Life Sciences (Wheaton) 358011 Potter-Elvehjem Safe-Grind Tissue Grinders, 30 mL WW-04468-34 Mfr # 358011
COMPARE
30 Glass 266
₹74,300.00INR / Case of 2