Cole-Parmer® Essentials Polystyrene Weighing Dishes - Anti-Static Square Weigh Boats
Rounded corners simplify transfer of powdered or granular samples
  • Disposable
  • White Translucent Color Dishes - for traditional applications
  • Blue Translucent Color Dishes - contrasts for white powders and solids, increases sample
General purpose sample holders. Translucent, rubberized polystyrene withstands temperatures to 180°F, flexes for easy pouring. Biologically inert; resistant to dilute acids, aqueous solutions, alcohols, bases. For use with balances, microwave moisture analyzers, hundreds of other sample handling applications. Flat bottom resists tipping. Contoured sides permit no-slip handling.
4,000.00 - ₹26,000.00INR / Pkg of 500
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Color Material
Availability Pricing
COMPARE
20 White Polystyrene
₹4,000.00INR / Pkg of 100
COMPARE
20 Blue Polystyrene
₹4,000.00INR / Pkg of 100
COMPARE
20 Blue Polystyrene
₹8,400.00INR / Pkg of 500
COMPARE
50 White Polystyrene
₹13,000.00INR / Case of 500
COMPARE
100 White Polystyrene
₹5,000.00INR / Pkg of 100
COMPARE
100 White Polystyrene
₹19,000.00INR / Pkg of 500
COMPARE
100 Blue Polystyrene
₹5,000.00INR / Pkg of 100
COMPARE
100 Blue Polystyrene
₹9,300.00INR / Pkg of 500
COMPARE
330 White Polystyrene
₹7,000.00INR / Pkg of 100
COMPARE
330 White Polystyrene
₹26,000.00INR / Pkg of 500
COMPARE
330 Blue Polystyrene
₹4,100.00INR / Pkg of 100
COMPARE
330 Blue Polystyrene
₹15,800.00INR / Pkg of 500
Showing 1- 12 of 12