Cole-Parmer® Essentials Polystyrene Weighing Dishes - Canoes with Pour Spout (Anti-Static)
Lets you pour weighed material without flexing dish—no more spills! Antistatic polystyrene
  • Disposable
These polystyrene weigh dish facilitates pouring and dispensing. No need to flex dish to dispense liquids or solids; 20-30% thicker material for strength, stability, handling ease, and safety. New weigh dishes have a flat bottom and will withstand 200°F. They are disposable, smooth, uniform, and inexpensive. Available in three convenient sizes for any sample handling application.
9,000.00 - ₹13,000.00INR / Pkg of 250
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Length (in) Width (in)
Availability Pricing
COMPARE
20 1 1/4 1 5/8
₹9,000.00INR / Pkg of 250
COMPARE
140 3 1/2 5 1/4
₹13,000.00INR / Pkg of 250
COMPARE
270 4 3/4 7 3/4
₹12,300.00INR / Pkg of 250