This product is now discontinued, View Related Products
PolyScience MX Series Digital Immersion Circulator
Converts any tank into a user-friendly circulating bath
  • Large user-friendly digital display
  • Integral heating coil and re-circulating pump
  • Adjustable mounting clamp
  • Built for laboratory and industrial use
This innovative immersion circulator is designed with a large, easy-to-read backlit display that shows both the process value and set value from a distance, as well as heat-on and circulating-on icons. The four tactile feedback push buttons provide simple control of the on/off/standby mode, set point value, Celsius/Fahrenheit selection, and offset calibration value. The low/high limit settings control the user-selectable min/max temperatures.
Compact, stylish design has a streamlined profile to occupy a minimum of reservoir space. A protective housing prevents direct contact with the integral pump and heater coil, while offering easy access for inspection and cleaning. A convenient slide control allows easy variable fluid flow adjustment. The adjustable mounting clamp attaches securely to flat and rounded tank walls. The single-speed pump effectively circulates liquids up to a 28-liter reservoir.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
PolyScience MXC-A11B Digital Immersion Circulator,28L, 120 VAC, 60 Hz WW-12101-61 Mfr # MX-CA11B
COMPARE
135 115 Digital Immersion Circulator,28L, 120 VAC, 60 Hz