This product is now discontinued, View Related Products

PolyScience Forced-Age Testing Bath (Beer Bath)

Ideal for accelerated shelf life testing of beer, wine and other beverages
  • Temperature range of -10 to 60°C
  • Reservoir capacity is 50 gallons (189 L)
Better known as the "beer bath," this bath is specifically designed for beer shelf life testing, food stability testing, product quality analysis, and asphalt sample tampering. The bath has a large open surface area, and hinged polycarbonate cover to optimize temperature stability.

Large 700 lbs (317.2 kg) bath must ship as freight transport.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC)
Availability Pricing
PolyScience Beer Bath Forced-Age Testing Bath; 208-230V WW-14575-05 Mfr # 070800
COMPARE
60 230
Call For Price
PolyScience Beer Bath Forced-Age Testing Bath; 240 VAC WW-14575-15 Mfr # 070805
COMPARE
50 230
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat