This product is now discontinued, View Related Products
PolyScience Benchtop and Compact Recirculating Chillers
Compact size maximizes bench space – simple operation and maintenance
  • Low noise output for quiet laboratory atmosphere
  • Top-mounted fill port with built-in fluid filter
  • Illuminated fluid-level indicator on front panel for easy viewing
These powerful chillers provide enough heat removal to simultaneously cool two rotary evaporators. Safety features include user-adjustable high-temperature limit and alarm auto-restart on power failure, and a fixed low flow rate alarm with power cutoff. Chiller features a closed 0.7 gal. (2.65 L) reservoir. Easy-access air filter keeps the chiller running at peak efficiency.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Cooling Capacity At 20° C (watts) Max Temperature (° C) Min Temperature (° C)
Availability Pricing
PolyScience LS51M11A110C Compact Recirculating Chiller, -20° to 40°C, 680W; 120 VAC WW-12930-10 Mfr # LS51M11A110C
COMPARE
1160 40 -20
Call For Price
PolyScience LS51MX1A110C Benchtop Recirculating Chiller, -20° to 40°C, 750W; 120 VAC WW-12930-30 Mfr # LS51MX1A110C
COMPARE
1290 40 -20
Call For Price
PolyScience LM61GX1A110C Benchtop Recirculating Chiller, -10° to 30°C, 350W; 120 VAC WW-12930-38 Mfr # LM61GX1A110C
COMPARE
560 30 -10