Polypropylene Utility Funnels
Get easy, economical funneling
  • Stackable
  • Dishwasher safe
  • Autoclavable.
These inexpensive and lightweight funnels feature side ribbing to let air escape when pouring samples. Ideal for general use and powders.
4,200.00 - ₹4,700.00INR / Pkg of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stem OD (mm) Top OD (mm)
Availability Pricing
COMPARE
70 10 70
₹4,200.00INR / Pkg of 12
COMPARE
125 8 83
₹4,500.00INR / Pkg of 12
COMPARE
300 8 83
₹4,700.00INR / Pkg of 12