Dynalon® Polyethylene Dippers
Lightweight, durable polyethylene (PE) dippers handle corrosives and liquid samples up to 248°F (120°C)
  • Handle end has fingergrip design for comfortable holding and a hole for hanging
The 36"L handles allow you to reach into large tanks. Dippers have securely welded solid handles and molded-in spouts for easy pouring.
9,000.00 - ₹15,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
Dynalon Polyethylene Dipper, 16 oz, 4" dia bowl, 36"L handle WW-06281-10 Mfr # 107075
COMPARE
500
₹9,000.00INR / Each
Dynalon Polyethylene Dipper, 32 oz, 5" dia bowl, 36"L handle WW-06281-50 Mfr # 106505
COMPARE
1000
₹15,600.00INR / Each