This product is now discontinued, View Related Products

Plastic Trans-Boxes Storage Containers

Snap-shurt covers retain contents even if overturned
Choose from three materials: transparent polystyrene (PS); sturdy, crystal-clear K-resin; or translucent, heavy-duty polypropylene (PP). Trans-Boxes containers with snap-shut covers are available in five configurations and three sizes.
Call For Price

13 variations of this product are available.

See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Number of Bins Description
Availability Pricing
COMPARE
12 Polystyrene container; 12 sections, 1-5/8\" x 3\" x 1-1/2\", 24/pack
Call For Price
COMPARE
6 Polystyrene container; 6 sections, 1-5/8\" x 6\" x 1-1/2\", 24/pack
Call For Price
COMPARE
12 Polystyrene container; 12 sections, 1-1/4\" x 2\" x 1-1/8\", 24/pack
Call For Price
COMPARE
6 Polystyrene container; 6 sections, 1-1/4\" x 4\" x 1-1/8\", 24/pack
Call For Price
COMPARE
1 Polystyrene container; 1 section, 8\" x 4\" x 1-3/16\", 24/pack
Call For Price
COMPARE
18 K-resin Storage Containers, 18 Sections, 1.6\" x 1.9\" x 1.5\"; 24/Pk
Call For Price
COMPARE
1 K-resin container; 1 section, 12-1/2\" x 8-1/4\" x 2\", 12/pack
Call For Price
COMPARE
6 K-resin container; 6 sections, 2\" x 8-1/4\" x 2\", 12/pack
Call For Price
COMPARE
12 K-resin container; 12 sections, 2\" x 4-1/8\" x 2\", 12/pack
Call For Price
COMPARE
24 K-resin container; 24 sections, 2\" x 2\" x 2\", 12/pack
Call For Price
COMPARE
6 K-resin container; 6 sections, 1-5/8\" x 6\" x 1-1/2\", 24/pack
Call For Price
COMPARE
6 Polypropylene container; 6 sections, 2\" x 8-1/4\" x 2\", 12/pack
Call For Price
COMPARE
24 Polypropylene container; 24 sections, 2\" x 2\" x 2\", 12/pack
Call For Price
Showing 1- 13 of 13

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat