Plastic Quick Disconnect Fittings, Barbed Inserts 16477
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
780.00 - ₹7,800.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material
Availability Pricing
CPC (Colder) APC22004 Quick-disconnect insert, acetal, 1/4" tubing ID, pack of 10 WW-06360-92 Mfr # APC22004
COMPARE
1/4 Acetal
₹1,500.00INR / Pkg of 10
CPC (Colder) APC22006 Quick-disconnect insert, acetal, 3/8" tubing ID, pack of 10 WW-06360-93 Mfr # APC22006
COMPARE
3/8 Acetal
₹780.00INR / Pkg of 10
CPC (Colder) MPC22004T03M Quick-disconnect fittings, Polycarbonate fittings, PP, 1/4" ID, insert, 5/pack WW-31311-61 Mfr # MPC22004T03M
COMPARE
1/4 Polycarbonate
₹1,300.00INR / Pkg of 5
CPC (Colder) MPC22006T03M Quick-disconnect fittings, Polycarbonate fittings, PP, 3/8" ID, insert, 5/pack WW-31311-62 Mfr # MPC22006T03M
COMPARE
3/8 Polycarbonate
₹1,300.00INR / Pkg of 5
CPC (Colder) MPX22803M Quick-disconnect fittings, Polycarbonate fittings, insert, 1/2" ID, 5/pack WW-31311-63 Mfr # MPX22803M
COMPARE
1/2 Polycarbonate
₹2,600.00INR / Pkg of 5
CPC (Colder) MPC22004T39M Quick-disconnect fittings, Polysulfone fittings, 1/4" ID, insert, 5/pack WW-31311-71 Mfr # MPC22004T39M
COMPARE
1/4 Polysulfone
₹2,500.00INR / Pkg of 5
CPC (Colder) MPC22006T39M Quick-disconnect fittings, Polysulfone fittings, 3/8" ID, insert, 5/pack WW-31311-72 Mfr # MPC22006T39M
COMPARE
3/8 Polysulfone
₹2,500.00INR / Pkg of 5
CPC (Colder) MPX22839M All-Plastic Quick Disconnect Barbed Inserts; Polysulfone (PS), 1/2" Tube ID, 5/pack WW-31311-73 Mfr # MPX22839M
COMPARE
1/2 Polysulfone
₹7,800.00INR / Pkg of 5
Showing 1- 8 of 8