Cole-Parmer® Plastic Buchner Funnels
Get quick, economical vacuum filtration
  • Easy sterilization: HDPE by gas, PP by autoclave or gas
Choose Buchner funnels for gravity or vacuum filtration. Recommended sterilization: HDPE for gas, PP by autoclave or gas.
2,300.00 - ₹33,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Paper Diameter (mm) Material Description
Availability Pricing
Dynalon High-density polyethylene Buchner funnel, 42.5 mm dia WW-06128-02 Mfr # 242835-042
COMPARE
42.5 High-density polyethylene (HDPE) High-density polyethylene Buchner funnel, 42.5 mm dia
₹2,300.00INR / Each
Dynalon High-density polyethylene Buchner funnel, 55 mm dia WW-06128-03 Mfr # 242835-055
COMPARE
55 High-density polyethylene (HDPE) High-density polyethylene Buchner funnel, 55 mm dia
₹2,800.00INR / Each
Dynalon Polypropylene Buchner funnel, 90 mm dia WW-06128-04 Mfr # 242835-090
COMPARE
90 Polypropylene (PP) Polypropylene Buchner funnel, 90 mm dia
₹5,400.00INR / Each
Dynalon Polypropylene Buchner funnel, 110 mm dia WW-06128-05 Mfr # 242835-110
COMPARE
110 Polypropylene (PP) Polypropylene Buchner funnel, 110 mm dia
₹7,500.00INR / Each
Dynalon Polypropylene Buchner funnel, 150 mm dia WW-06128-10 Mfr # 242835-150
COMPARE
150 Polypropylene (PP) Polypropylene Buchner funnel, 150 mm dia
₹13,400.00INR / Each
Dynalon Polypropylene Buchner funnel, 240 mm dia WW-06128-20 Mfr # 242835-240
COMPARE
240 Polypropylene (PP) Polypropylene Buchner funnel, 240 mm dia
₹33,000.00INR / Each