This product is now discontinued, View Related Products

Pelican Unbreakable Instrument Cases

Protect your valuable instrumentation – cases are absolutely air- and water-tight
  • Customize your case with prescored “Pick-N-Pluck” foam
  • Cases are made of unbreakable, corrosion-proof polypropylene
  • Built-in O-ring keeps case air- and water-tight
  • (Instruments are not included)
All Pelican cases are constructed of high-impact polypropylene and feature ABS plastic latches. A ¼" thick neoprene O-ring guarantees a water- and dust-proof seal. Each case includes a "Pick-N-Pluck” foam liner — customize your case by simply removing pieces of foam. Models 40460-92 and -94 also include adjustable handle and rolling wheels for easy transport.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Exterior Height (in) Exterior Length (in) Exterior Width (in)
Availability Pricing
Pelican 1120-BLK Unbreakable Instrument Case, 7-3/8" x 4-7/8" x 3-9/16" Interior WW-40460-78 Mfr # 1120-BLK
COMPARE
3 9/16 7 3/8 4 7/8
Call For Price
Pelican 1150-000-110 Unbreakable Instrument Case, 8-1/2" x 6" x 3-3/4" Interior WW-40460-80 Mfr # 1150-000-110
COMPARE
4 9 7
Call For Price
Pelican 1200-000-110 Unbreakable Instrument Case, 9-1/2" x 7-1/2" x 4-1/2" Interior WW-40460-82 Mfr # 1200-000-110
COMPARE
5 10 9
Call For Price
Pelican 1400-000-110 Unbreakable Instrument Case, 12-1/4" x 9-1/4" x 5-1/4" Interior WW-40460-84 Mfr # 1400-000-110
COMPARE
6 13 12
Call For Price
Pelican 1500-000-110 Unbreakable Instrument Case, 17" x 11-3/4" x 6" Interior WW-40460-86 Mfr # 1500-000-110
COMPARE
6 7/8 18 15
Call For Price
Pelican 1600-000-110 Unbreakable Instrument Case, 22" x 17" x 8" Interior WW-40460-88 Mfr # 1600-000-110
COMPARE
9 24 19
Call For Price
Pelican 1620-BLK Unbreakable Instrument Caseinterior Case 22-1/16" x 17" x 12-9/16" WW-40460-92 Mfr # 1620-BLK
COMPARE
13 3/4 24 3/4 19 1/2
Call For Price
Pelican 1650-BLK Unbreakable Instrument Caseinterior Case 29" x 17-7/8" x 10-1/2" WW-40460-94 Mfr # 1650-BLK
COMPARE
11 1/4 32 1/5 20 1/2
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat