PEEK High-Pressure Unions, Tees, and Crosses
Suitable for use with high pressure chromatography applications
  • Constructed of chemically inert PEEK plastic
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16" OD tubing.
6,900.00 - ₹15,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
high-pressure unions, tees, and crosses; 0.020\" bore, union
₹6,900.00INR / Each
COMPARE
high-pressure unions, tees, and crosses; 0.050\" bore, union
₹6,900.00INR / Each
COMPARE
high-pressure unions, tees, and crosses; 0.020\" bore, tee
₹7,600.00INR / Each
COMPARE
high-pressure unions, tees, and crosses; 0.050\" bore, cross
₹15,600.00INR / Each