PEAK Scientific Solaris 10 Air Compressors
Support Solaris 10 nitrogen generator for labs without an external high quality air supply
  • Stackable with Solaris 10 nitrogen generators
  • Engineered for ELSD and Compact Mass Spectroscopy
  • Compact, stackable design to maximize the use of your lab space
  • Available in 110 and 230 VAC
Solaris air compressor is recommended to support the Solaris 10 nitrogen generator, providing an on-site gas supply of high-purity nitrogen. It can be placed on a benchtop and stacked with the nitrogen generator on top to contain gas supply in a single system while maximizing your lab space.
Solaris generators are the most advanced nitrogen generator on the market today – designed from the ground up to deliver maximum performance for labs that cannot afford to compromise. By design, Solaris XE is simple to set up and use. However, as an added benefit, installation and training services are offered where dedicated Peak field service engineers will visit you to assist with getting you up and running with no stress – order 86443-41 in addition to your generator.
1,300,000.00 - ₹1,455,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (bar) Power (VAC) Description
Availability Pricing
PEAK Scientific Solaris Air Compressor for Solaris 10; 230 VAC WW-86443-08 Mfr # 65-0003
COMPARE
8.27 230 Air Compressor for Solaris 10; 230 VAC
₹1,300,000.00INR / Each
PEAK Scientific Solaris Air Compressor for Solaris 10; 110 VAC WW-86443-09 Mfr # 65-0004
COMPARE
8.27 110 Air Compressor for Solaris 10; 110 VAC
₹1,455,000.00INR / Each