PCRmax® Systemic Viral Pathogen Detection Kits for qPCR
Quickly screen for viral pathogens to prevent the spreading of disease causing systemic viruses
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Prevent economic loss, especially in the dairy and agricultural industry, using these viral detection kits to accurately assess whether these disease-causing pathogens are present. Select a complete kit with mastermix or order a kit without mastermix and use your own.
100 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
No RNA African Horse Sickness Virus, without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA African Horse Sickness Virus, with mastermix
₹214,000.00INR / Each
COMPARE
No DNA Aleutian Disease Virus, without mastermix
₹164,000.00INR / Each
COMPARE
Yes DNA Aleutian Disease Virus, with mastermix
₹209,000.00INR / Each
COMPARE
No RNA Avian Infectious Bronchitis Virus (IBV), without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Avian Infectious Bronchitis Virus (IBV), with mastermix
₹236,000.00INR / Each
COMPARE
No RNA Avian Influenza A Virus Subtype H5, without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Avian Influenza A Virus Subtype H5, with mastermix
₹262,000.00INR / Each
COMPARE
No RNA Avian Influenza A Virus Subtype H7, without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Avian Influenza A Virus Subtype H7, with mastermix
₹214,000.00INR / Each
COMPARE
No RNA Avian Influenza A Virus Subtype H7N9, without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Avian Influenza A Virus Subtype H7N9, with mastermix
₹262,000.00INR / Each
COMPARE
No RNA Avian Influenza A Virus Subtype H9, without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Avian Influenza A Virus Subtype H9, with mastermix
₹214,000.00INR / Each
COMPARE
No RNA Avian Orthoreovirus, without mastermix
₹242,000.00INR / Each
COMPARE
Yes RNA Avian Orthoreovirus, with mastermix
₹292,000.00INR / Each
COMPARE
No DNA Beak and Feather Disease Virus, without mastermix
₹152,000.00INR / Each
COMPARE
Yes DNA Beak and Feather Disease Virus, with mastermix
₹212,000.00INR / Each
COMPARE
No RNA Bird Flu, without mastermix
₹159,000.00INR / Each
COMPARE
Yes RNA Bird Flu, with mastermix
₹254,000.00INR / Each
Showing 1 - 20 of 100