PCRmax® Systemic Microbial Pathogen Detection Kits for qPCR
Screen for systemic microbial pathogens quickly to prevent economic loss
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Prevent economic loss, especially in the dairy and agricultural industry, using these microbial detection kits to accurately assess whether these disease-causing pathogens are present. Select a complete kit with mastermix or order a kit without mastermix and use your own.
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
Yes DNA Salmonella (all pathogenic species), with mastermix
₹169,000.00INR / Each
COMPARE
Yes DNA Alpha toxin producing Clostridium perfringens, with mastermix
₹272,000.00INR / Each
COMPARE
Yes DNA Aspergillus fumigatus, with mastermix
₹142,000.00INR / Each
COMPARE
Yes DNA Candidatus Mycoplasma haemominutum, with mastermix
₹272,000.00INR / Each
COMPARE
Yes DNA Enterocytozoon bieneusi, with mastermix
₹153,000.00INR / Each
COMPARE
Yes DNA Giardia intestinalis (human infections), with mastermix
₹170,000.00INR / Each
COMPARE
No DNA Listeria monocytogenes, without mastermix
₹132,000.00INR / Each
COMPARE
Yes DNA Mycoplasma felis, with mastermix
₹164,000.00INR / Each
Showing 1- 8 of 8