This product is now discontinued, View Related Products
PCRmax® qPCR Plates
Improve your heat transfer and cycle times
  • Constructed of high-quality polypropylene for optimal clarity and solvent resistance
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Suitable for use in most thermal cyclers
  • Choose from 96- and 384-well plates
The qPCR plates feature ultrathin walls for accelerated heat transfer and cycle times. The 96-well plates are available with either 0.15- or 0.2-mL well volume; skirted, half-skirted, or unskirted; and low or standard profile well. The 384-well plate features a 40 µl well volume, 25 µl sample volume, and is skirted. The plates are designed to fit most major thermal cyclers and are autoclavable.
24,700.00 - ₹87,500.00INR / Case of 50
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Number of Wells Well Volume (mL) Description
Availability Pricing
COMPARE
96 0.2 qPCR Plate 96-Well white, low profile, full skirt, 50/cs
₹36,800.00INR / Case of 50
COMPARE
96 0.2 qPCR Plate 96-Well white, standard profile, half skirt, 50/cs
₹24,700.00INR / Case of 50
COMPARE
96 0.2 qPCR Plate 96-Well white, low profile, half skirt, 50/cs
₹24,700.00INR / Case of 50
COMPARE
96 0.2 qPCR Plate 96-Well white, low profile, half skirt; raised rim, 50/cs
₹24,700.00INR / Case of 50
COMPARE
96 0.2 qPCR Plate 96-Well white, standard profile, no skirt, 50/cs
₹24,700.00INR / Case of 50
COMPARE
96 0.2 qPCR Plate 96-Well white, low profile, no skirt, 50/cs
₹24,700.00INR / Case of 50
COMPARE
96 0.15 qPCR Plate 96-Well white for RLC 480, low profile, half skirted, 50/cs
Call For Price
COMPARE
96 0.15 qPCR Plate 96-Well white for RLC 480, low profile, half skirted & self-adhesive sealing film, 50/cs
₹87,500.00INR / Case of 50
COMPARE
384 0.04 qPCR Plate 384-Well white, full skirted, 50/cs
₹51,400.00INR / Case of 50
COMPARE
Call For Price
Showing 1- 10 of 10