This product is now discontinued, View Related Products
PCRmax PCR Transparent Plates
Ultrathin walls provide faster heat transfer and cycle times
  • Constructed of high-quality polypropylene for optimal clarity and solvent resistance
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Choose from 96- and 384-well plates
The transparent 96- and 384-well plates are designed with ultra-thin walls offering accelerated heat transfer and quicker cycle times. The 96-well plates have a well volume of 0.2 mL, are available with full, half, or no skirt, and come with a low or standard profile well. The 384-well plate has a well volume of 40 µl, a sample volume of 25 µl, and features a full skirt. The plates are compatible with most thermal cyclers and are autoclavable.
24,700.00INR / Case of 50
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Number of Wells Well Volume (mL) Description
Availability Pricing
COMPARE
96 0.2 PCR Plate 96-Well clear, low profile, full skirt, 50/cs
Call For Price
COMPARE
96 0.2 PCR Plate 96-Well clear, standard profile, half skirt, 50/cs
Call For Price
COMPARE
96 0.2 PCR Plate 96-Well clear, low profile, half skirt, 50/cs
₹24,700.00INR / Case of 50
COMPARE
96 0.2 PCR Plate 96-Well clear, low profile, half skirt, raised rim, 50/cs
₹24,700.00INR / Case of 50
COMPARE
96 0.2 PCR Plate 96-Well clear, standard profile, no nskirt, 50/cs
₹24,700.00INR / Case of 50
COMPARE
96 0.2 PCR Plate 96-Well clear, low profile, no skirt, 50/cs
₹24,700.00INR / Case of 50
COMPARE
384 0.4 PCR Plate 384-Well clear, full skirted, 50/cs
Call For Price
COMPARE
384 0.4 PCR Plate 384-Well clear, full skirted, 50/cs
Call For Price
Showing 1- 8 of 8