This product is now discontinued, View Related Products
PCRmax PCR Strip Tube Caps
  • Constructed of pure high-quality polypropylene without additives
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
  • Available in five color options
Choose between domed and flat-style caps in five color options: clear, blue, red, yellow, or green.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Cap Style Tube Color Description
Availability Pricing
COMPARE
Domed Clear PCR Strip Tube Caps, Domed, Clear; 125/Cs
Call For Price
COMPARE
Domed Red PCR Strip Tube Caps, Domed, Red; 125/Cs
Call For Price
COMPARE
Domed Yellow PCR Strip Tube Caps, Domed, Yellow; 125/Cs
Call For Price
COMPARE
Domed Green PCR Strip Tube Caps, Domed, Green; 125/Cs
Call For Price
COMPARE
Domed Blue PCR Strip Tube Caps, Domed, Blue; 125/Cs
Call For Price
COMPARE
Flat Clear PCR/qPCR Strip Tube Caps, Flat, Clear; 125/Cs