PCRmax® Human Microbial Pathogen Detection Kits for qPCR
Quickly and accurately detect the presence of human microbial pathogens
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
Yes DNA Enteropathogenic Escherichia coli, with mastermix
₹153,000.00INR / Each
COMPARE
Yes DNA Klebsiella oxytoca, with mastermix
₹222,000.00INR / Each
COMPARE
No DNA Pseudomonas aeruginosa, without mastermix
₹120,000.00INR / Each
COMPARE
Yes DNA Pseudomonas aeruginosa, with mastermix
₹153,000.00INR / Each
COMPARE
Yes DNA Staphylococcus aureus, with mastermix
₹153,000.00INR / Each
COMPARE
Yes DNA Staphylococcus epidermidis, with mastermix
₹153,000.00INR / Each
COMPARE
Yes DNA DNA 150 test lyophilised OneStep qRT-PCR Mastermix
₹51,300.00INR / Each
COMPARE
Yes DNA QPCR KIT QPCR MASTERMIX
₹92,300.00INR / Each
Showing 1- 8 of 8