Cole-Parmer® DLF-200 Series PCR Verticular Laminar Flow Cabinets
Protect your PCR samples against cross-contamination
  • Built-in germicidal UV lamp permits decontamination
  • Verticular laminar airflow with HEPA filtration
  • Innovative filtration technology
  • Produces ISO Class 5 compliant clean air
  • Meets OSHA, ANSI, and other international standards
The Cole-Parmer ductless PCR Worksation is the perfect cabinet for nonhazardous applications where biological or biohazard by-products are not generated, and user protection is not required. The cabinet provides a contaminant-free interior with a built-in germicidal 253.7 nm UV lamp. The lamp has a timer to permit decontamination between cycles—set timer up to 60 minutes or to hold indefinitely.
Innovative filtration technology consists of a pre-filter and main HEPA filter which is 99.99% effective for 0.3 micron particulates. The filter chamber prevents contaminated air from encountering internal cabinet components, prolonging the lifespan. A filter door lock prevents unauthorized removal or accidental exposure to dirty filters.
529,000.00 - ₹766,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Blower Type Interior Width (in) Power (VAC)
Availability Pricing
COMPARE
ebm-papst centrifugal fan 24 115
₹543,000.00INR / Each
COMPARE
ebm-papst centrifugal fan 24 230
₹529,000.00INR / Each
COMPARE
ebm-papst centrifugal fan 36 115
₹673,000.00INR / Each
COMPARE
ebm-papst centrifugal fan 36 230
₹673,000.00INR / Each
COMPARE
ebm-papst centrifugal fan 48 115
₹766,000.00INR / Each
COMPARE
ebm-papst centrifugal fan 48 230
₹766,000.00INR / Each