Monarch Instrument PalmStrobe x Portable Stroboscopes
Removable, rechargeable battery pack allows unlimited measuring power
  • Compact, lightweight housing allows for convenient one-handed operation
  • Four-quadrant tuner makes stop action and slow motion inspections easy
The PalmStrobe x portable stroboscope is the industrial standard for high intensity, multifunction xenon stroboscopes. Memory holds eight commonly used flash patterns and the last flash rate used - no need to recalculate flash rates each time. Memory is retained even when the battery is recharging - recharge battery in or out of strobe.
The 6-digit, backlit LCD display is easy to read in both well-lit or dim locations. Additional features include rapid x2 and ÷2 of current flash rate, TTL-compatible input/output, and low-battery indicator.
18,800.00 - ₹192,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Display Type Flash Duration
Availability Pricing
Monarch 6205-050 PalmStrobe x, with 115/230 VAC recharger WW-53510-00 Mfr # 6205-050
COMPARE
6-digit alphanumeric backlit LCD, 0.3\" H 10 to 30 µsec typical
Call For Price
Monarch 6205-051 PalmStrobe x Pak, with 115/230 VAC recharger, spare battery and holster WW-53510-01 Mfr # 6205-051
COMPARE
6-digit alphanumeric backlit LCD, 0.3\" H 10 to 30 µsec typical
Call For Price
Monarch 6205-052 s PalmStrobe x Stroboscope Kit WW-53510-02 Mfr # 6205-052
COMPARE
6-digit alphanumeric backlit LCD, 0.3\" H 10 to 30 µsec typical
₹192,000.00INR / Each
Monarch 6280-026 Rechargeable NiMH battery pack for Palm Strobe x WW-53510-04 Mfr # 6280-026
COMPARE
Call For Price
Monarch 6280-028 Replacement Xenon Lamps for Stroboscopes, L-1905; 2/Pk WW-53510-05 Mfr # 6280-028
COMPARE
₹18,800.00INR / Pkg of 2
Monarch Instrument 6280-037 PalmStrobe x Portable Stroboscope Pulse Output Cable WW-98806-98 Mfr # 6280-037
COMPARE
Call For Price
Monarch 6280-042 PalmStrobe x CC-9 Portable Stroboscope Latching Plastic Case WW-98807-00 Mfr # 6280-042
COMPARE
Call For Price
Showing 1- 7 of 7