This product is now discontinued, View Related Products
Omnifit® SolventPlus™ Chromatography Columns with One Adjustable and One Fixed Endpieces
Get the highest chemical compatibility
  • Exceptional chemical resistance with wetted parts of PTFE, ETFE, borosilicate glass, and Chemraz®
  • Adjustable endpiece offers quick and easy height adjustment
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
These SolventPlus columns are designed for techniques using organic solvents such as reversed phase chromatography, but are equally compatible with aqueous solvent systems. The most inert materials are present in the fluid path to provide superior chemical resistance. Use the columns in low pressure or gravity feed applications.
Columns come with one fixed and one adjustable endpieces pre-assembled with 30 µm PTFE frits. Adjustable endpiece gives you the flexibility to modify the bed length up to 80 mm in height plus the ability to make fine adjustments while causing minimal disturbance to the packed bed. Each column is supplied assembled and ready to use, with all the tubing, fittings, and connections needed to get started.
71,300.00 - ₹271,000.00INR / Each
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Column Inside Diameter (ID) (mm) Column Length (mm)
Availability Pricing
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 50 mm; 1/ea WW-11946-01 Mfr # 006SCC-06-05-AF
COMPARE
0.1 to 0.7 6.6 50
₹71,300.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 100 mm; 1/ea WW-11946-04 Mfr # 006SCC-06-10-AF
COMPARE
0.1 to 2.4 6.6 100
₹71,700.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 150 mm; 1/ea WW-11946-07 Mfr # 006SCC-06-15-AF
COMPARE
1.4 to 4.1 6.6 150
₹73,200.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 250 mm; 1/ea WW-11946-10 Mfr # 006SCC-06-25-AF
COMPARE
4.8 to 7.5 6.6 250
₹75,700.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 400 mm; 1/ea WW-11946-13 Mfr # 006SCC-06-40-AF
COMPARE
9.9 to 12.7 6.6 400
₹79,400.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 100 mm; 1/ea WW-11946-16 Mfr # 006SCC-10-10-AF
COMPARE
0.2 to 5.5 10 100
₹81,400.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 150 mm; 1/ea WW-11946-19 Mfr # 006SCC-10-15-AF
COMPARE
3.1 to 9.4 10 150
₹82,500.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 250 mm; 1/ea WW-11946-22 Mfr # 006SCC-10-25-AF
COMPARE
11.0 to 17.3 10 250
₹85,400.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 400 mm; 1/ea WW-11946-25 Mfr # 006SCC-10-40-AF
COMPARE
22.8 to 29.1 10 400
₹88,900.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 500 mm; 1/ea WW-11946-28 Mfr # 006SCC-10-50-AF
COMPARE
30.6 to 36.9 10 500
₹92,100.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 100 mm; 1/ea WW-11946-31 Mfr # 006SCC-15-10-AF
COMPARE
0.4 to 12.4 15 100
₹109,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 150 mm; 1/ea WW-11946-34 Mfr # 006SCC-15-15-AF
COMPARE
7.1 to 21.2 15 150
₹110,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 250 mm; 1/ea WW-11946-37 Mfr # 006SCC-15-25-AF
COMPARE
24.7 to 38.9 15 250
₹112,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 400 mm; 1/ea WW-11946-40 Mfr # 006SCC-15-40-AF
COMPARE
51.2 to 65.4 15 400
₹113,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 500 mm; 1/ea WW-11946-43 Mfr # 006SCC-15-50-AF
COMPARE
68.9 to 83.0 15 500
₹110,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 100 mm; 1/ea WW-11946-46 Mfr # 006SCC-25-10-AF
COMPARE
1.2 to 34.4 25 100
₹123,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 150 mm; 1/ea WW-11946-49 Mfr # 006SCC-25-15-AF
COMPARE
19.6 to 58.9 25 150
₹130,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 250 mm; 1/ea WW-11946-52 Mfr # 006SCC-25-25-AF
COMPARE
68.7 to 181.6 25 250
₹127,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 400 mm; 1/ea WW-11946-55 Mfr # 006SCC-25-40-AF
COMPARE
142.3 to 181.6 25 400
₹135,000.00INR / Each
Diba Omnifit® SolventPlus™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 500 mm; 1/ea WW-11946-58 Mfr # 006SCC-25-50-AF
COMPARE
191.4 to 230.7 25 500
₹131,000.00INR / Each
Showing 1 - 20 of 26