Diba Omnifit® Packing Sleeves for EZ and EZ SolventPlus™ Columns
Use packing sleeve to join two columns for filling and packing
  • Suitable for use with Diba Omnifit® EZ and EZ SolventPlus™ columns
These Diba Omnifit® packing sleeves allow you to easily join two columns or connect a column to a column packer. Choose from a wide range of column inner diameter sizes suitable for your application.
6,400.00 - ₹33,400.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm)
Availability Pricing
Diba Omnifit® EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 5 mm; 1/ea WW-11942-14 Mfr # 006EPS-05
COMPARE
5
₹6,400.00INR / Each
Diba Omnifit® EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 6.6 mm; 1/ea WW-11942-15 Mfr # 006EPS-06
COMPARE
6.6
₹6,400.00INR / Each
Diba Omnifit® EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 10 mm; 1/ea WW-11942-16 Mfr # 006EPS-10
COMPARE
10
₹8,700.00INR / Each
Diba Omnifit® EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 15 mm; 1/ea WW-11942-17 Mfr # 006EPS-15
COMPARE
15
₹12,100.00INR / Each
Diba Omnifit® EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 25 mm; 1/ea WW-11942-18 Mfr # 006EPS-25
COMPARE
25
₹12,100.00INR / Each
Diba Omnifit® EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 35 mm; 1/ea WW-11942-19 Mfr # 006EPS-35
COMPARE
35
₹16,600.00INR / Each
Diba Omnifit® EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 50 mm; 1/ea WW-11942-20 Mfr # 006EPS-50
COMPARE
50
₹33,400.00INR / Each
Showing 1- 7 of 7